Dissolved oxygen bottles
Oxygen bottle acc. to Winkler, uncalibrated, without numbering, 100 ml, 19/26
Katalog # 705-0570
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Bottle, approx. 100 ml capacity, made from clear soda lime glass with grounded glass stopper. Height with stopper approx. 115 to 120 mm. Accessories include a clamp that holds the stopper in place.
Specifikationer
- Kapacitet:100 ml
- Beskrivelse:Oxygen bottle acc. to Winkler, uncalibrated, without numbering
- NS prop:19/26
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Stopper holder for dissolved oxygen bottlesStopper holder for dissolved oxygen bottles705-0573Log ind for tilgængelighed1 * 1 Items - 72,90 kr