Orders LinkContactUsLinkComponent
Accessories for dropping bottles
Accessories for dropping bottles
Supplier:   NORDIC PACK
Accessories for dropping bottles
Supplier:  NORDIC PACK
Certificates
Accessories for dropping bottles
Accessories for dropping bottles
2 Options Available Below
Product Details & Documents
Cap, pipette, and rubber suction bulb, available as spare parts for brown dropping bottles.
Product Family Options
Product Information
  • Beskrivelse
    Til
    Availability
    Price
  • Screw cap, pipette and rubber suction bulb
    10 ml dropping bottles
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 33,50 kr
    33,50 kr
    1 * 1 Items
       33,50 kr
  • Screw cap, pipette and rubber suction bulb
    25 ml dropping bottles
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 18,20 kr
    18,20 kr
    1 * 1 Items
       18,20 kr