VWR® Short Thread Vials ND9
Short thread vial with integrated micro-insert, 0,20 ml, 11,6×32,0 mm, Glas, ravfarvet
Katalog # 548-2649A
Leverandør: VWR Collection
Gå til:
Product Details & Documents
- 1st hydrol. class, writing patch and filling lines, with small micro-insert
- TopBond
Documents
CoC VWR Short Thread Vials ND9.pdf
Overensstemmelseserklæring
Specifikationer
- Diameter×H:11,6×32,0 mm
- Kapacitet:0,20 ml
- Farve:ravfarvet
- Beskrivelse:Short thread vial with integrated micro-insert
- Materiale:Glas