Orders LinkContactUsLinkComponent
VWR® Cleanroom Stool
VWR® cleanroom stool, 450 - 650 mm, Castors
Katalog # 132-1452
Leverandør:   VWR Collection
VWR® Cleanroom Stool
Katalog # 132-1452
Leverandør:  VWR Collection
VWR® Cleanroom Stool
VWR® Cleanroom Stool
VWR® cleanroom stool, 450 - 650 mm, Castors
Price
3.910,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This VWR® stool is suitable for clean rooms, certified and complete ESD safety. Featuring an easy and convenient height adjustment from any sitting position thanks to a ring release underneath the seat, which is protected from contamination by the soft cover.
  • 450 – 650 mm height adjustment
  • 400 mm seat diameter
  • Air cleanliness class 3 according to DIN EN ISO 14644-1
  • Exceeds the ESD requirements of DIN EN 61340-5-1
  • GS certificate for "tested safety"
The extra-large seat is made of synthetic black leather upholstery (60 mm high) and the rounded front edge of the seat prevents possible blood congestion in the thighs. The integral foam upholstery offers excellent sitting comfort, maximum resistance, and ease of cleaning. Extremely robust, durable, washable, disinfectant-proof, PVC-free, resistant to blood and urine, fire protection in accordance with EN 1021 Parts 1 + 2. The seat automatically encourages a correct sitting posture and upright, healthy sitting.
The particularly small aluminium base with a diameter of 540 mm is space-saving and allows high mobility due to the available legroom. The large, specially designed laboratory castors (Ø 65 mm) , move smoothly and are easy to clean. The stool is braked when not used which decreases the risk of accidents. This cleanroom stool meets standard DIN 67 877, weighs only 7 kg and is environmentally friendly as all materials can be recycled separately.
Warranty: 10-year long-term guarantee, free shipping of spare parts during the whole period of validity.
Specifikationer
  • Beskrivelse:
    VWR® cleanroom stool
  • Højde:
    450 - 650 mm
  • Størrelse:
    Castors