Orders LinkContactUsLinkComponent
Airlock Tray Handle
Airlock Tray Handle
Airlock tray handle
Katalog # DOWIA09422
Leverandør:   DON WHITLEY SCIENTIFIC
Airlock Tray Handle
Katalog # DOWIA09422
Leverandør:  DON WHITLEY SCIENTIFIC
Airlock Tray Handle
Airlock tray handle
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
A useful handle for use with Whitley airlock trays. This tool gives you additional reach to pull trays from the airlock, move them around inside the workstation and push trays back into the airlock.
Specifikationer
  • Beskrivelse:
    Airlock tray handle