Spare Filter Foam
Spare filiter foam
Katalog # IKAA20113275
Leverandør: IKA
Gå til:
Product Details & Documents
Spare filter foam for covers of the multiDrive milling chambers. For possible replacement after multiple cleaning of the filter foam.
- Width: 50 mm
- Height: 10 mm
Leveringsinformation: Scope of delivery: 5 pieces/pack.
Specifikationer
- Beskrivelse:Spare filiter foam