Classroom DNA electrophoresis LabStation™
Catalog # 732-1421
Supplier: EDVOTEK
Specifications
- Beskrivelse:DNA LabStation™
- Pakn:1
Specifications
About this item
This LabStation provides all that is needed to run any Edvotek DNA or dye electrophoresis kits with students.
Delivery: System includes M36 HexaGel™ electrophoresis tank (six 7×7 cm trays), DuoSource™ power supply (75 V for one or two units), fixed volume micropipette (40 µl), yellow micropipette tips (1 to 200 µl/2 racks of 96), and a DNA Fingerprinting Classroom Kit.