ContactUsLinkComponent
Experimentation kits, molecular weight determination of proteins (agarose-based)
Experimentation kits, molecular weight determination of proteins (agarose-based)
Katalog # EDVO110
Leverandør:  EDVOTEK
undefined
Experimentation kits, molecular weight determination of proteins (agarose-based)
Katalog # EDVO110
Leverandør:  EDVOTEK
Specifikationer
  • Beskrivelse:
    Molecular Weight Determination of Proteins (Agarose-based)
  • Pakn:
    1 KIT
Specifikationer
Om denne vare
This experiment introduces a simple method to determine protein subunit molecular weights using horizontal electrophoresis. As the protein standards and 'unknowns' are prestained, the separation of proteins can be observed during electrophoresis. Included in the experiment is protein-grade agarose, which provides an alternative to the use of polyacrylamide gels.
  • For 4 lab groups
  • Complete in 1 hour followed by overnight staining
Also required: Horizontal electrophoresis apparatus, power supply, white light visualisation system, 5 to 50 µl adjustable or 20 µl fixed volume micropipette, methanol, and glacial acetic acid.
Delivery: Kit includes instructions, prestained LyphoProtein samples, gel loading solution, agarose, electrophoresis buffer, stain, and SDS solution.