Ready-to-Load™ kits, the case of the invisible bands
Catalog # 763-0009
Supplier: EDVOTEK
Specifications
- Beskrivelse:The Case of the Invisible Bands
- Pakn:1
Specifications
About this item
Experiment using fluorescence and electrophoresis to solve the mystery of the invisible bands.
- For 10 lab groups
- Complete in 45 minutes
Also required: Electrophoresis tank, power supply and long wave UV lamp.
Delivery: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.