Orders LinkContactUsLinkComponent
PCR beads
PCR beads
PCR Beads, 1
Katalog # EDVO625
Leverandør:   EDVOTEK
PCR beads
Katalog # EDVO625
Leverandør:  EDVOTEK
CAS nummer:  
PCR beads
PCR Beads, 1
CAS nummer:  
Price
494,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Each PCR Bead contains Taq DNA polymerase, Taq DNA polymerase buffer, dNTP mixture and MgCl₂. Pack contains sufficient for 25 reactions.
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    PCR Beads