Orders LinkContactUsLinkComponent
DNA Staining System, FlashBlue™, Edvotek
DNA Staining System, FlashBlue™, Edvotek
FlashBlue™ DNA Staining System, 60 mL
Katalog # EDVO609
Leverandør:   EDVOTEK
DNA Staining System, FlashBlue™, Edvotek
Katalog # EDVO609
Leverandør:  EDVOTEK
CAS nummer:  
DNA Staining System, FlashBlue™, Edvotek
FlashBlue™ DNA Staining System, 60 mL
CAS nummer:  
Price
135,00 kr
Log ind for tilgængelighed
1 * 60 mL
Product Details & Documents
FlashBlue™ is a DNA stain that offers simple and rapid staining of agarose gels.
  • 10X Concentrate for 1,2 L
Concentrated liquid that, when diluted, can be used for both rapid and overnight staining of DNA fragments.
Specifikationer
Specifikationer
  • Pakn:
    60 mL
  • Beskrivelse:
    FlashBlue™ DNA Staining System
Specifikationer