Experimentation kits, Mini-Prep isolation of plasmid DNA
Catalog # EDVO202
Supplier: EDVOTEK
Specifications
- Beskrivelse:Mini-Prep Isolation of Plasmid DNA
- Pakn:1 KIT
Specifications
About this item
Small-scale, rapid isolation of plasmid DNA is a routine procedure used for screening and analysis of recombinant DNAs in cloning and sub-cloning experiments. In this experiment, students isolate plasmid DNA without the use of toxic chemicals such as phenol or chloroform.
- For 5 lab groups
- Complete in 60 minutes
Also required: Electrophoresis tank and power supply, water batch, microcentrifuge (10000 min⁻¹), 5 to 50 µl adjustable or 40 and 50 µl fixed volume micropipettes, 95 to 100% isopropanol, and white light box.
Delivery: Kit includes instructions, various solutions and buffers, agarose powder, and stains.