Coloured beads
Farvede perler
Katalog # EDVO1500
Leverandør: EDVOTEK
Gå til:
Product Details & Documents
A set of coloured beads that can be designated to represent the Watson-Crick DNA bases: A, T, G, C.
Can be used to demonstrate concepts related to the structure and biology of DNA.
Bestillingsoplysninger: Includes 150 beads of each colour with detailed outline of various demonstrations.
Specifikationer
- Beskrivelse:Farvede perler