Orders LinkContactUsLinkComponent
VWR® Vacuum Aspiration Systems
VWR® Vacuum Aspiration Systems
VWR® Vacuum Aspiration Systems
VWR® Vacuum Aspiration Systems
The VWR® vacuum aspiration system is designed for laboratory waste recovery and the separation of liquids and solids.
  • Compact, space-saving, user-friendly design
  • Level detection system prevents bottle overfill (4 L)
  • Parts in contact with liquids are autoclavable
  • Contamination and protect personnel safety
  • Adjustable vacuum level with indicator
  • Collection bottle outlet with hydrophobic filter 0,45 μm prevents aerosol and liquid contamination
  • 15 ml/s aspiration rate
Designed with compact and space-saving housing for laboratory waste recovery and the separation of liquids and solids.
Package includes main unit, one bottle, one operating handle with tube, one single-needle, one long type needle, one 8-channel-needle, one tip detrusion adapter, and one 8-channel injector.
 : EMC and LVD, UKCA certificated.
 : Two-year warranty.
 : VWR® 4 L and 2 L vacuum aspiration system basic unit is supplied with main unit, 4 L bottle for 4 L version and 2 L bottle for 2 L version, handle set with eight kinds of standard adapters, hydrophobic filter 0,45 μm. Components are also available separately.
  • Catalog No:
  • 181-0659
  • 181-0658
  • Flaskekapacitet:
  • 2 L
  • 4 L
  • Pump flow rate:
  • 15 L/min (air)
  • 15 L/min (air)
  • Vakuumområde:
  • 0 - 600 mbar (adjustable)
  • 0 - 600 mbar (adjustable)
  • Vægt:
  • 2,8 kg (6,2 lbs.)
  • 3,0 kg (6,6 lbs.)
Product Family Options
  • Beskrivelse
    B×D×H
  • Vacuum aspiration system, 2 L
    180×240×340 mm
    1 * 1 Items
    1 * 1 Items - 3.410,00 kr
    3.410,00 kr
    1 * 1 Items
       3.410,00 kr
    Specifications
    Cat. No.
    181-0659
    Flaskekapacitet
    2 L
    Pump flow rate
    15 L/min (air)
    Vakuumområde
    0 - 600 mbar (adjustable)
    Vægt
    2,8 kg (6,2 lbs.)
  • Vacuum aspiration system, 4 L
    180×240×450 mm
    1 * 1 Items
    1 * 1 Items - 6.720,00 kr
    6.720,00 kr
    1 * 1 Items
       6.720,00 kr
    Specifications
    Cat. No.
    181-0658
    Flaskekapacitet
    4 L
    Pump flow rate
    15 L/min (air)
    Vakuumområde
    0 - 600 mbar (adjustable)
    Vægt
    3,0 kg (6,6 lbs.)
Accessories
  •  
    Beskrivelse
  • 181-0664
    Tip detrusion adapter,4L
    1 * 1 Items - 63,50 kr
  •  
    Beskrivelse
  • 181-0671
    Eight-channel needle Ø 1,5 mm for 4 and 2 L
    1 * 1 Items - 112,00 kr
  •  
    Beskrivelse
  • 181-0661
    4 L PPCO vacuum bottle
    1 * 1 Items - 584,00 kr
  •  
    Beskrivelse
  • 181-0663
    Hydrophobic filter 0,45 μm
    1 * 1 Items - 190,00 kr
  •  
    Beskrivelse
  • 181-0668
    Single needle Ø 2,5 mm, 40 mm for 4 and 2 L
    1 * 1 Items - 32,90 kr
  •  
    Beskrivelse
  • 181-0666
    Single needle Ø 1,5 mm, 40 mm for 4 and 2 L
    1 * 1 Items - 32,90 kr
  •  
    Beskrivelse
  • 181-0670
    Eight-channel tip detrusion adapter 200 μl tip for 4 and 2 L
    1 * 1 Items - 47,80 kr
  •  
    Beskrivelse
  • 181-0660
    VWR® Non-contact liquid level sensor
    1 * 1 Items - 791,00 kr
  •  
    Beskrivelse
  • 181-0669
    Single needle Ø 2,5 mm, 120 mm for 4 and 2 L
    1 * 1 Items - 32,90 kr
  •  
    Beskrivelse
  • 181-0662
    2 L vacuum bottle
    1 * 1 Items - 347,00 kr
  •  
    Beskrivelse
  • 181-0665
    Tip detrusion adapter, 2 L
    1 * 1 Items - 63,50 kr
  •  
    Beskrivelse
  • 181-0667
    Single needle Ø 1,5 mm, 120 mm for 4 and 2 L
    1 * 1 Items - 32,90 kr