HLP Medium (Hsu's Lactobacillus/Pediococcus Medium)
HLP Medium (Hsu's Lactobacillus/Pediococcus Medium), 500 g, Plastic bottle, 1
Leverandør: DOEHLER GROUP
HLP Medium (Hsu's Lactobacillus/Pediococcus Medium)
Katalog # DOEH923556244
Leverandør: DOEHLER GROUP
CAS nummer:
Gå til:
Product Details & Documents
Siebel-Instituts nutrient medium for the selective detection and quantitative determination of Lactobacillus and Pediococcus from the brewing environment.
- Certified by the Siebel-Institute for technology
- Ready-to-use powder to prepare HLP gel or agar without prior autoclaving
- Functionally tested with strains originally isolated from the brewing environment
HLP promote the growth of beer spoiling bacteria like Lactobacillus spp./Pedicococcus spp. which are commonly encountered in a brewery. Cycloheximide contained in the medium inhibits the growth of culture yeast.
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Plastic bottle
- Pakn:1
- Beskrivelse:HLP Medium (Hsu's Lactobacillus/Pediococcus Medium)
- Påfyldning:500 g