Orders LinkContactUsLinkComponent
SteriWare Cake Cutters
SteriWare Cake Cutters
Leverandør:   Sampling Systems
SteriWare Cake Cutters
Leverandør:  Sampling Systems
Certifikater
SteriWare Cake Cutters
SteriWare Cake Cutters
2 tilgængelige muligheder nedenfor
Product Details & Documents
These circular cutters are perfect for taking samples of cake like material.
  • Made from FDA and EC compliant HDPE
  • 100% cleanroom manufactured and packed
  • Choice of sterile and non sterile
These individually bagged samplers are perfect for taking small 2 to 3 cm samples of cake-like material typically from in trays.
Product Family Options
Non sterile
Produktinformation
  • Materiale
    Længde
    Tilgængelighed
    Price
  • SAMP8765H-4050
    SteriWare cake cutter, 40 mm, non sterile
    SteriWare cake cutter, 40 mm, non sterile
    HDPE
    50 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 Items
    1 * 50 Items - 950,00 kr
    950,00 kr
    1 * 50 Items
       950,00 kr
Sterile
Produktinformation
  • Materiale
    Længde
    Tilgængelighed
    Price
  • SAMP8765H-4050S
    SteriWare cake cutter, 40 mm, sterile
    SteriWare cake cutter, 40 mm, sterile
    HDPE
    50 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 Items
    1 * 50 Items - 1.300,00 kr
    1.300,00 kr
    1 * 50 Items
       1.300,00 kr