Experimentation kits, analysis and comparision of mammalian cell types
Katalog # EDVO986
Leverandør: EDVOTEK
Specifikationer
- Beskrivelse:Analysis and Comparision of Mammalian Cell Types
- Pakn:1
Specifikationer
Om denne vare
Observe the differences between mammalian cell types and how these cells funciton. Cells are fixed on microscope slides and students stain the cells on the slide to view morphological characteristics of the cell types. These cells are very safe for classroom use.
- For 6 lab groups
- Complete in 35 minutes
- Requires a microscope with 400x magnification
Delivery: Kit includes instructions, multispot slides (4 cell types each), eosin and methylene blue stain, mounting medium, cover slips, transfer pipettes, and immersion troughs.