Heat resistant gloves, Clavies® biohazard autoclave gloves, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
These soft, pliant, all-cotton terry cloth gloves are superb for handling hot objects out of the autoclave or oven, or for other heat protection in the lab, school or industry. Shield hands and forearms when unloading the autoclave.
- Bright orange colour for immediate identification and visibility
- Heat-resistant to 232 °C
- Available with longer length cuff (28 cm), ideal for use in larger autoclaves
- Terry cloth construction can be machine washed and tumble dried
Specifications
- Catalog No:
- 112-1991
- 112-1990
- Model:
- Biohazard Autoclave
- Biohazard Autoclave
- Ambidextrous/Hånd-specifik:
- Hånd-specifik
- Hånd-specifik
- Engangs/Genbrugelig:
- Genanvendelig
- Genanvendelig
- Manchet:
- Lang tyk
- Lang tyk
- Cuff længde:
- 28 cm
- 13 cm