Sterile Pipette Filter Tips in Rack
Catalog # KART0963500
Supplier: KARTELL
Specifications
- Pakket:10 racks, 96 each
- Steril:+
- Volumen:2 - 100 µl
- Længde:50 mm
- Spidstype:Low retention; graduated
- Spits farge:Natural
- Til brug med:Kartell® PL100, Gilson®, Eppendorf ®, Biohit®, Socorex®, HTL®, Nichiryo®, Finnpipette®, Thermo®
- Filter:+
- Pakn:10
Specifications
About this item
These tips are made of high quality, 100% pure, virgin PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
- Sterilised by gamma radiation
- Designed for use in molecular biology, cell cultures, bacteriology, virology, immunology and radioimmunology
- Hydrophobic pe filters act as a barrier against aerosol contamination, preventing cross-contamination of samples and micropipettes
- Tip orifices are sized to completely empty the tip in the event of over-aspiration
Made of PP and including a filter, these tips are DNase, RNase and DNA free, ATP and metal free, cadmiuim free, apyrogenic and PCR inhibitor free.