Orders LinkContactUsLinkComponent
Cerex® HPSCX SPE Columns
Cerex® HPSCX SPE Columns
Leverandør:   Tecan
Cerex® HPSCX SPE Columns
Leverandør:  Tecan
Certifikater
Cerex® HPSCX SPE Columns
2 tilgængelige muligheder nedenfor
Product Details & Documents
Mixed mode sorbent with both reverse-phase and strong cationic exchange chemistries.
  • Ion-exchange
  • Liquid handler compatible
Its high surface area provides excellent capacity and a high selectivity for basic analytes. Ideal for drug monitoring, screening, identification, confirmation and quantitation.
Product Family Options
Produktinformation
  • Fase
    Partikelstørrelse
    Kapacitet
    Lejevægt
    Tilgængelighed
    Price
  • TCAT427-0353C
    Column Cerex HPSCX
    Column Cerex HPSCX
    HPSCX
    30 µm
    3 ml
    35 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 Items
    1 * 100 Items
  • TCAT427-0506C
    Column Cerex HPSCX
    Column Cerex HPSCX
    HPSCX
    30 µm
    6 ml
    50 mg