Orders LinkContactUsLinkComponent
Cerex® PSAXS SPE Columns
Cerex® PSAXS SPE Columns
Leverandør:   Tecan
Cerex® PSAXS SPE Columns
Leverandør:  Tecan
Certifikater
Cerex® PSAXS SPE Columns
2 tilgængelige muligheder nedenfor
Product Details & Documents
Mixed mode reverse-phase with an anion exchanger for acidic compounds.
  • Ion-exchange
  • Liquid handler compatible
A 10 µm silica micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
Product Family Options
Produktinformation
  • Fase
    Porestørrelse
    Partikelstørrelse
    Kapacitet
    Lejevægt
    Tilgængelighed
    Price
  • TCAT606-0501R
    Column Cerex PSAXS
    Column Cerex PSAXS
    PSAX-S
    300 Å
    10 µm
    1 ml
    50 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 96 Items
    1 * 96 Items
  • TCAT606-0503C
    Column Cerex PSAXS
    Column Cerex PSAXS
    PSAX-S
    300 Å
    10 µm
    3 ml
    50 mg