Orders LinkContactUsLinkComponent
Resolvex® M10 Manual Positive Pressure Processor
Leverandør:   Tecan
Resolvex® M10 Manual Positive Pressure Processor
Leverandør:  Tecan
Certifikater
Resolvex® M10 Manual Positive Pressure Processor
3 tilgængelige muligheder nedenfor
Product Details & Documents
Benchtop manual positive pressure processor.
  • Individual flow restrictors ensure parallel processing even when some wells are vacant
  • Only a gas supply is needed for operation
  • Dual-level gas delivery system offers fine controls of flow and pressure
Perfect for entry into positive pressure processing and workflows, the intuitive design of the Resolvex® M10 makes it easy to use with minimal training required.
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • TCAT289-0004
    Resolvex® M10 48 instrument, 355×305×390 mm
    Resolvex® M10 48 instrument, 355×305×390 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • TCAT288-0001
    Resolvex® M10 96 1 ml/NBE columns instrument, 259×305×390 mm
    Resolvex® M10 96 1 ml/NBE columns instrument, 259×305×390 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
  • TCAT288-0006
    Resolvex® M10 96-XT 96 1 ml/NBE columns instrument, 259×305×390 mm
    Resolvex® M10 96-XT 96 1 ml/NBE columns instrument, 259×305×390 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • SPE column rack for 6 ml columns for 48 place, aluminum, W×D×H: 10,8×3,6×3,5"
    SPE column rack for 6 ml columns for 48 place, aluminum, W×D×H: 10,8×3,6×3,5"
    TCAT279-0023
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • SPE column rack for 1 ml cerex columns, aluminum, W×D×H: 10,8×3,6×3,1"
    SPE column rack for 1 ml cerex columns, aluminum, W×D×H: 10,8×3,6×3,1"
    TCAT279-0021
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • SPE column rack for 3 ml columns for 48 place, aluminum, W×D×H: 10,8×3,6×3,5"
    SPE column rack for 3 ml columns for 48 place, aluminum, W×D×H: 10,8×3,6×3,5"
    TCAT279-0022
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Partial plate holder for 1 ml columns (generic), aluminum, W×D×H: 3,1×3,4×5,0"
    Partial plate holder for 1 ml columns (generic), aluminum, W×D×H: 3,1×3,4×5,0"
    TCAT253-0006
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Waste barrier insert (long), plastic, W×D×H: 4,4×3,0×1,6"
    Waste barrier insert (long), plastic, W×D×H: 4,4×3,0×1,6"
    TCAT253-0036-SP
    Log ind for tilgængelighed
    1 * 5 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Collection rack for 12×75 mm tubes for 48 place system, stainless steel, W×D×H: 10,7×3,5×3,8"
    Collection rack for 12×75 mm tubes for 48 place system, stainless steel, W×D×H: 10,7×3,5×3,8"
    TCAT279-0015
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Partial plate holder for NBE columns (generic), aluminum, W×D×H: 3,1×3,4×5,0"
    Partial plate holder for NBE columns (generic), aluminum, W×D×H: 3,1×3,4×5,0"
    TCAT253-0007
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Collection rack for 16×100 tubes for 48 mm place system, stainless steel, W×D×H: 10,7×3,5×3,8"
    Collection rack for 16×100 tubes for 48 mm place system, stainless steel, W×D×H: 10,7×3,5×3,8"
    TCAT279-0016
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Collection rack for 12×32 mm autosampler vials for 48 place systems, stainless steel, W×D×H: 10,7×3,5×3,8"
    Collection rack for 12×32 mm autosampler vials for 48 place systems, stainless steel, W×D×H: 10,7×3,5×3,8"
    TCAT279-0017
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Mat Luer seal pull tab silicone, H=5,11 mm
    Mat Luer seal pull tab silicone, H=5,11 mm
    TCAT253-2003
    Log ind for tilgængelighed
    1 * 5 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Collection rack to fit 12×32 mm autosampler vials, stainless steel, W×D×H: 10,7×3,5×3,9"
    Collection rack to fit 12×32 mm autosampler vials, stainless steel, W×D×H: 10,7×3,5×3,9"
    TCAT279-0018
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Reservoir 96 well 300 ml pyramid, plastic, W×D×H: 5,0×3,4×1,7"
    Reservoir 96 well 300 ml pyramid, plastic, W×D×H: 5,0×3,4×1,7"
    TCAT700-4009
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Mat 96 well pre-slit silic BlankSilicone, H=5,11 mm
    Mat 96 well pre-slit silic BlankSilicone, H=5,11 mm
    TCAT278-0060
    Log ind for tilgængelighed
    1 * 10 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Plate V-bottom solid 5 ml, plastic, W×D×H: 5,0×3,4×0,6"
    Plate V-bottom solid 5 ml, plastic, W×D×H: 5,0×3,4×0,6"
    TCAT700-4008
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Rack sample 6CC 20 mm spacing, plastic, W×D×H: 5,0×3,4×2,9"
    Rack sample 6CC 20 mm spacing, plastic, W×D×H: 5,0×3,4×2,9"
    TCAT30219826
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Rack sample 3CC 20 mm spacing, plastic, W×D×H: 5,0×3,4×2,9"
    Rack sample 3CC 20 mm spacing, plastic, W×D×H: 5,0×3,4×2,9"
    TCAT30219827
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Rack collection tube 12×75 mm assay, plastic, W×D×H: 5,0×3,6×3,2"
    Rack collection tube 12×75 mm assay, plastic, W×D×H: 5,0×3,6×3,2"
    TCAT30219825
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Rack vial autosampler assay, plastic, W×D×H: 5,3×3,6×1,7"
    Rack vial autosampler assay, plastic, W×D×H: 5,3×3,6×1,7"
    TCAT30219821
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Rack MN giga assay, plastic, W×D×H: 5,0×3,4×4,4"
    Rack MN giga assay, plastic, W×D×H: 5,0×3,4×4,4"
    TCAT30219819
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Rack QIAGEN-tip mega assay, plastic, W×D×H: 5,0×3,4×4,3"
    Rack QIAGEN-tip mega assay, plastic, W×D×H: 5,0×3,4×4,3"
    TCAT30219818