Orders LinkContactUsLinkComponent
TBX agar
TBX agar
TBX agar, 90 mm ready-to-use plates, -, 20
Katalog # LIOF10522
Leverandør:   LIOFILCHEM
TBX agar
Katalog # LIOF10522
Leverandør:  LIOFILCHEM
CAS nummer:  
TBX agar
TBX agar, 90 mm ready-to-use plates, -, 20
CAS nummer:  
Price
262,00 kr
Log ind for tilgængelighed
1 * 20 Items
Product Details & Documents
X-glucuronide, contained in the TBX agar formulation, is the chromogenic agent that allows the determination of the β-Dglucuronidase activity, which is a highly specific enzyme for E. coli. Gram-positive bacteria are inhibited by bile salts.
Specifikationer
Specifikationer
  • Pakn:
    20
  • Beskrivelse:
    TBX agar, 90 mm ready-to-use plates
  • Påfyldning:
Specifikationer