Orders LinkContactUsLinkComponent
Cleanroom Cubby Racks
Cleanroom Cubby Racks
48 space racks available with 7,5"/190 mm slot space
Catalog # PALBGC1302
Supplier:   Palbam
Cleanroom Cubby Racks
Catalog # PALBGC1302
Supplier:  Palbam
Cleanroom Cubby Racks
48 space racks available with 7,5"/190 mm slot space
Price
71.410,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Cleanroom cubby racks.
  • Stainless steel 304L
  • Electro-polished
  • On leveler feet
  • With lip at front
Specifications
  • Beskrivelse:
    48 space racks available with 7,5"/190 mm slot space