HIGH STAT PLUS ESD Jackets
Leverandør: ALSICO HIGHTECH
Om denne vare
Unisex ESD jackets.
- Upright collar
- Centre front closure with hidden zipper and press studs
- Adjustable waistband with press studs
- Chest pocket left with zipper
- Chest pocket right with flap and hidden press studs
- 2 side pockets with vertical zipper
- Inset sleeves
- Adjustable sleeve ends with press studs
- Accents in royal blue
- ESD logo
Industrial launderable.
: Oeko-Tex® standard 100; IEC 61340 - 4 - 9:2016.
6 tilgængelige muligheder nedenfor
Product Details & Documents
Unisex ESD jackets.
- Upright collar
- Centre front closure with hidden zipper and press studs
- Adjustable waistband with press studs
- Chest pocket left with zipper
- Chest pocket right with flap and hidden press studs
- 2 side pockets with vertical zipper
- Inset sleeves
- Adjustable sleeve ends with press studs
- Accents in royal blue
- ESD logo
Industrial launderable.
: Oeko-Tex® standard 100; IEC 61340 - 4 - 9:2016.
Specifications for Product Family
Specifikationer
- Catalog No:
- ALSO1017456-XS
- ALSO1017456-S
- ALSO1017456-M
- ALSO1017456-L
- ALSO1017456-XL
- ALSO1017456-2XL
- Outfit applikationer:
- Cleanroom|Industrial
- Cleanroom|Industrial
- Cleanroom|Industrial
- Cleanroom|Industrial
- Cleanroom|Industrial
- Cleanroom|Industrial
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Køn:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Stof:
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- Vægt:
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
Specifikationer
Product Family Options
Produktinformation
- ModelFarveTøj størrelseBeklædningsgenstandTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.ALSO1017456-XSOutfit applikationerCleanroom|IndustrialRisk areaESDKønUnisexSleeve or leg lengthLongStof68% polyester/30,5% cotton/1,5% carbonVægt175 g/m² - Specifikationer:
More
Specifications
Cat. No.ALSO1017456-SOutfit applikationerCleanroom|IndustrialRisk areaESDKønUnisexSleeve or leg lengthLongStof68% polyester/30,5% cotton/1,5% carbonVægt175 g/m² - Specifikationer:
More
Specifications
Cat. No.ALSO1017456-MOutfit applikationerCleanroom|IndustrialRisk areaESDKønUnisexSleeve or leg lengthLongStof68% polyester/30,5% cotton/1,5% carbonVægt175 g/m² - Specifikationer:
More
Specifications
Cat. No.ALSO1017456-LOutfit applikationerCleanroom|IndustrialRisk areaESDKønUnisexSleeve or leg lengthLongStof68% polyester/30,5% cotton/1,5% carbonVægt175 g/m² - Specifikationer:
More
Specifications
Cat. No.ALSO1017456-XLOutfit applikationerCleanroom|IndustrialRisk areaESDKønUnisexSleeve or leg lengthLongStof68% polyester/30,5% cotton/1,5% carbonVægt175 g/m² - Specifikationer:
More
Specifications
Cat. No.ALSO1017456-2XLOutfit applikationerCleanroom|IndustrialRisk areaESDKønUnisexSleeve or leg lengthLongStof68% polyester/30,5% cotton/1,5% carbonVægt175 g/m²