Orders LinkContactUsLinkComponent
LPS for Endotoxin Indicator
LPS for Endotoxin Indicator
LPS for Endotoxin indicator, with endotoxin level >100,000 EU/vial, 6
Katalog # WAKFWLPS-0100K
Leverandør:   FUJIFILM WAKO CHEMICALS
LPS for Endotoxin Indicator
Katalog # WAKFWLPS-0100K
CAS nummer:  
LPS for Endotoxin Indicator
LPS for Endotoxin Indicator
LPS for Endotoxin indicator, with endotoxin level >100,000 EU/vial, 6
CAS nummer:  
Price
3.820,00 kr
Log ind for tilgængelighed
1 * 6 Items
Product Details & Documents
The LPS for Endotoxin Indicator is composed of E. coli O55:B5 (>100,000 EU/vial) and serves as a stock solution to prepare multiple endotoxin indicators for depyrogenation validation studies.
  • E. coli 055:B5 (>100,000 EU/vial)
  • Can make multiple endotoxin indicators from one vial
  • Used for dry heat oven validation studies
The prepared endotoxin indicators may be tested using reagents from our PYROSTAR™ ES-F series.
Documents
Specifikationer
Specifikationer
  • Pakn:
    6
  • Beskrivelse:
    LPS for Endotoxin indicator, with endotoxin level >100,000 EU/vial
Specifikationer