Executive Series™ Housekeeping Carts
The Executive Compact housekeeping cart is a complete system solution for efficient housekeeping.
- Lightweight and ergnonoic design allows for easy maneuverability and optimum comfort
- Impact-absorbing and non-marring bumpers are designed to reduce costly wall damage
- Full-size cabinet offers exceptional storage space and an adjustable middle shelf
- Multiple accessories available for a customised solution
Double bag capacity allows for multiple collection streams such as linens and waste or recycling with watertight PVC lining.
Delivery information: Medium housekeeping cart includes canvas bag with vinyl lining, bumpers, vacuum holder, bungee kit. Large housekeeping cart includes canvas bag with vinyl lining, bumpers, vacuum holder, and bungee kit. High security housekeeping cart includes 2 canvas bags with vinyl lining, 2 sanitizing caddies 9 L, security compartment, and locking cabinet kit.
Documents
TDS Large Housekeeping Cart
Datablad
- Farve:sort
- Beskrivelse:High security housekeeping cart
- Seq:03
- B×D×H:131,4×55,9×135,9 mm
- Vægt:35,8 kg
- Cat. No.:129-1945
- Materiale:Plast
Accessories
- Beskrivelse
- 129-2085Vinyl bag, brown, collects up to 128 L of waste (20% more than traditional cart bags) with zippered front for easy waste removal, for carts RUBB1805985, RUBB1860740, RUBBFG9T7200BLA, RUBBFG9T7400BLA, RUBBFG9T7500BLA, RUBBFG9T7800BLA1 * 1 Items - 954,00 kr
- Beskrivelse
- 129-2086Vinyl bag, blue, collects up to 128 L of waste (20% more than traditional cart bags) with zippered front for easy waste removal, for carts RUBB1805985, RUBB1860740, RUBBFG9T7200BLA, RUBBFG9T7400BLA, RUBBFG9T7500BLA, RUBBFG9T7800BLA1 * 1 Items - 954,00 kr
- Beskrivelse
- 129-1959Waste cover with storage compartment, plastic, black, covers waste collection while providing storage for shopping centre items like pens, check lists, and other items, for carts RUBB1805985, RUBB1860740, RUBBFG9T7200BLA, RUBBFG9T7400BLA, RUBBFG9T7500BLA, RUBBFG9T7800BLA1 * 1 Items - 512,00 kr
- Beskrivelse
- 129-1984Locking door kit, plastic, black, secures and conceals supplies and amenities throughout the housekeeping process, for carts RUBBFG618900BLA, RUBBFG619000BLA, RUBBFG9T7800BLA1 * 1 Items - 1.530,00 kr
- Beskrivelse
- 129-1944Set of 2×9 L caddies; provides a quick and easy way to clean in tight places, yellow, plastic, for carts RUBBFG9T7200BLA, RUBBFG9T7500BLA, RUBBFG9T7800BLA1 * 1 Items - 296,00 kr
- Beskrivelse
- 129-2087Vinyl bag, yellow, collects up to 128 L of waste (20% more than traditional cart bags) with zippered front for easy waste removal, for carts RUBB1805985, RUBB1860740, RUBBFG9T7200BLA, RUBBFG9T7400BLA, RUBBFG9T7500BLA, RUBBFG9T7800BLA1 * 1 Items - 954,00 kr