Orders LinkContactUsLinkComponent
Serological Pipettes Holder
Serological Pipettes Holder
Holder for serological pipettes, 300×100×420 mm
Katalog # KART0111500
Leverandør:   KARTELL
Serological Pipettes Holder
Katalog # KART0111500
Leverandør:  KARTELL
Serological Pipettes Holder
Serological Pipettes Holder
Holder for serological pipettes, 300×100×420 mm
Price
2.990,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Bench pipette holder, made of particularly clear polymethylmethacrylate (PMMA), has high level finishes and its content can be seen from any angle.
  • Complete with four tilted support shelves and white top
The four separate compartments are ideal for housing plastic or glass serologic pipettes of various capacities (from 1 to 50 ml). Do not shine ultraviolet light on it.
Specifikationer
  • Beskrivelse:
    Holder for serological pipettes
  • B×D×H:
    300×100×420 mm