Orders LinkContactUsLinkComponent
Mop frame, Swep Duo Plus
Mop frame, Swep Duo Plus
Swep Duo ramme, 50 cm
Katalog # 129-0457
Leverandør:   FHP VILEDA SCS
Mop frame, Swep Duo Plus
Katalog # 129-0457
Leverandør:  FHP VILEDA SCS
Mop frame, Swep Duo Plus
Swep Duo ramme, 50 cm
Price
354,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The double sided Swep Duo mop frame lets you clean twice the area by just flipping the frame and keep mopping. Swep Duo Plus frame with built-in lever is designed for touchless removal of dirty and/or contaminated mops.
  • Lightweight and ergonomic
  • Trapezoidal shape for easy cleaning of corners
  • Works with all core and specialist application double and single sided mops
Recommended handle: Aluminium telescopic handle (119966).
Specifikationer
  • Beskrivelse:
    Swep Duo ramme
  • Længde:
    50 cm
  • Seq:
    06