Orders LinkContactUsLinkComponent
Bis(2-ethylhexyl)adipate
Bis(2-ethylhexyl)adipate
200 L
Catalog # SPCMD1246-200LTBL
Irritant
Supplier:   Spectrum Chemical
Bis(2-ethylhexyl)adipate
Catalog # SPCMD1246-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  103-23-1
Bis(2-ethylhexyl)adipate
200 L
CAS Number:  103-23-1
Price
20.320,00 kr
Log in for availability
1 * 200 L
Product Details & Documents
Dioctyl Adipate is a diester which is used as a plasticizer.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Synonyms:  Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate
  • CAS Number:103-23-1
  • Formula:C₂₂H₄₂O₄
  • MDL Number:MFCD00009496
  • Molecular Weight:370.57 g/mol
  • Density:0,927
  • Storage Temperature:Køleskab
  • EINECS:203-090-1
Specifications
Specifications
  • Pakn:
    200 L