Orders LinkContactUsLinkComponent
Dimethyl glutarate ≥98%
Dimethyl glutarate ≥98%
200 L
Katalog # SPCMD2948-200LTBL
Leverandør:   Spectrum Chemical
Dimethyl glutarate ≥98%
Katalog # SPCMD2948-200LTBL
Leverandør:  Spectrum Chemical
CAS nummer:  1119-40-0
Dimethyl glutarate ≥98%
200 L
CAS nummer:  1119-40-0
Price
32.420,00 kr
Log ind for tilgængelighed
1 * 200 L
Product Details & Documents
Dimethyl Glutarate is used in de-icing and anti-icing applications, construction and road applications, and release and binders agents.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Synonymer:  1,5-Pentanedioic acid dimethyl ester, Glutaric acid dimethyl ester
  • CAS nummer:1119-40-0
  • Formel:C₇H₁₂O₄
  • MDL nummer:MFCD00008468
  • Molecular Weight:160.17 g/mol
  • Merck Index:13,04486
  • Kogende Pt:211 °C (1013 hPa)
  • Smeltende Pt:–37 °C
  • Tæthed:1,087
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:214-277-2
Specifikationer
Specifikationstestresultater
  • Assay:
    98% Min.
Specifikationer
  • Pakn:
    200 L