Orders LinkContactUsLinkComponent
Nitrotyrosine IP Kit
Nitrotyrosine IP Kit
Kit contains: Affinity sorbent, antibody, BSA, spin columns, and collection tubes, 1
Katalog # CAYM601220-1
Leverandør:   Cayman Chemical
Nitrotyrosine IP Kit
Katalog # CAYM601220-1
Leverandør:  Cayman Chemical
CAS nummer:  
Nitrotyrosine IP Kit
Nitrotyrosine IP Kit
Kit contains: Affinity sorbent, antibody, BSA, spin columns, and collection tubes, 1
CAS nummer:  
Price
5.530,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
For the capture and pulldown of nitrated proteins from cell and tissue lysates.
  • Determine change in tyrosine nitration in treated versus non-treated samples
  • Demonstrate target proteins are nitrated in vitro
  • Identify and characterise nitrated proteins by Western blot and proteomic analysis
Cayman’s Nitrotyrosine IP Kit allows for the capture and pulldown of nitrated proteins from cell lysates using a sorbent coupled with a nitrotyrosine monoclonal antibody. The nitrated proteins can be eluted from the resin, loaded to an SDS-PAGE gel, and probed using the included Nitrotyrosine Polyclonal Antibody or a user supplied target specific antibody. A positive control sample assures that the assay is working.
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    Kit contains: Affinity sorbent, antibody, BSA, spin columns, and collection tubes
  • Cat. No.:
    CAYM601220-1
  • Varemærke:
    CAYM