Imhoff Cone without Stopcock
Imhoff cone, scale up to 1000 ml, ring mark at 1000 ml, Without stopcock
Catalog # HECH44275010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Made from borosilicate 3.3 glass.
- Very good chemical resistance
- Excellent thermal resistance
- With frosted label and scale
- Fused white graduation
The sedimentation cone with graduation according to Imhoff is used for the quantification of sediments from a suspension.
Certifications: Produced according to DIN 12672.
Specifications
- Model:Without stopcock
- Beskrivelse:Imhoff cone, scale up to 1000 ml, ring mark at 1000 ml