Orders LinkContactUsLinkComponent
Jenway® pH Electrodes
Jenway® pH Electrodes
Leverandør:   Cole-Parmer
Jenway® pH Electrodes
Leverandør:  Cole-Parmer
Certifikater
Jenway® pH Electrodes
9 tilgængelige muligheder nedenfor
Product Details & Documents
A variety of pH electrodes for your Jewnay meters.
  • Choose the right electrode for your application
These electrodes are designed for standard measurements of general solutions. The epoxy body makes it especially suitable for field measurements or use in education.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • JENW99960-41
  • JENW99960-34
  • JENW99960-39
  • JENW68006-37
  • JENW90269-17
  • JENW68006-40
  • JENW99960-44
  • JENW59530-70
  • JENW79000-71
  • Model:
  • pH
  • pH
  • pH
  • pH
  • pH
  • pH
  • pH
  • pH
  • pH
  • Kabellængde:
  • 1 m
  • 1 m
  • 1 m
  • 1 m
  • 1 m
  • 1 m
  • 1 m
  • 1 m
  • 1 m
  • pH-område:
  • 0 - 14 pH
  • 0 - 14 pH
  • 0 - 14 pH
  • 0 - 14 pH
  • 0 - 14 pH
  • 0 - 14 pH
  • 0 - 14 pH
  • 0 - 14 pH
  • 0 - 14 pH
  • Reference electrolyte:
  • AgCl, liquid filled single junction
  • AgCl, gel filled single junction
  • Double junction Ag/AgCl
  • AgCl
  • AgCl, liquid filled single junction
Specifikationer
Product Family Options
Produktinformation
  • Model
    Temperatur
    Materiale
    Ø×L
    Tilslutning
    Til
    Tilgængelighed
    Price
  • JENW99960-41
    Life science refillable combination with bulb end
    Life science refillable combination with bulb end
    pH
    0...80 °C
    Glas
    12×120 mm
    BNC
    Jenway® pH meters
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.630,00 kr
    1.630,00 kr
    1 * 1 Items
       1.630,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    JENW99960-41
    Model
    pH
    Kabellængde
    1 m
    pH-område
    0 - 14 pH
    Reference electrolyte
    AgCl, liquid filled single junction
  • JENW99960-34
    General purpose epoxy bodied pH electrode (without temperature compensation)
    General purpose epoxy bodied pH electrode (without temperature compensation)
    pH
    0...80 °C
    Epoxy
    12×120 mm
    BNC
    Any pH meter with BNC connection
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 876,00 kr
    876,00 kr
    1 * 1 Items
       876,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    JENW99960-34
    Model
    pH
    Kabellængde
    1 m
    pH-område
    0 - 14 pH
    Reference electrolyte
    AgCl, gel filled single junction
  • JENW99960-39
    Refillable combination with bulb end type
    Refillable combination with bulb end type
    pH
    0...50 °C
    Glas
    12×120 mm
    BNC
    Jenway® pH meters
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.300,00 kr
    2.300,00 kr
    1 * 1 Items
       2.300,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    JENW99960-39
    Model
    pH
    Kabellængde
    1 m
    pH-område
    0 - 14 pH
    Reference electrolyte
    Double junction Ag/AgCl
  • JENW68006-37
    Refillable reference with bulb end
    Refillable reference with bulb end
    pH
    0...50 °C
    Glas
    12×120 mm
    2 mm plug
    Jenway® pH meters
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.850,00 kr
    1.850,00 kr
    1 * 1 Items
       1.850,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    JENW68006-37
    Model
    pH
    Kabellængde
    1 m
    pH-område
    0 - 14 pH
    Reference electrolyte
  • JENW90269-17
    Refillable spear type combination with bulb end
    Refillable spear type combination with bulb end
    pH
    0...80 °C
    Glas
    12×120 mm
    BNC
    Jenway® pH meters
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.610,00 kr
    1.610,00 kr
    1 * 1 Items
       1.610,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    JENW90269-17
    Model
    pH
    Kabellængde
    1 m
    pH-område
    0 - 14 pH
    Reference electrolyte
  • JENW68006-40
    Sealed combination with bulb end and protective cover
    Sealed combination with bulb end and protective cover
    pH
    0...60 °C
    Epoxy
    12×120 mm
    S7
    Jenway® pH meters
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 995,00 kr
    995,00 kr
    1 * 1 Items
       995,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    JENW68006-40
    Model
    pH
    Kabellængde
    1 m
    pH-område
    0 - 14 pH
    Reference electrolyte
  • JENW99960-44
    Sealed combination with bulb end and protective cover
    Sealed combination with bulb end and protective cover
    pH
    0...80 °C
    Epoxy
    12×120 mm
    3150/3205 waterproof connector
    Jenway® pH meters
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.970,00 kr
    2.970,00 kr
    1 * 1 Items
       2.970,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    JENW99960-44
    Model
    pH
    Kabellængde
    1 m
    pH-område
    0 - 14 pH
    Reference electrolyte
  • JENW59530-70
    Waterproof, general purpose, epoxy-bodied pH electrode with ATC temperature compensation
    Waterproof, general purpose, epoxy-bodied pH electrode with ATC temperature compensation
    pH
    0...100 °C
    Epoxy
    12×120 mm
    3150/3205 waterproof connector
    Jenway® 3150 and 3205 pH meters
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.880,00 kr
    1.880,00 kr
    1 * 1 Items
       1.880,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    JENW59530-70
    Model
    pH
    Kabellængde
    1 m
    pH-område
    0 - 14 pH
    Reference electrolyte
    AgCl
  • JENW79000-71
    12 mm stem spear type pH electrode, glass bodied for measurement of soil and slurry samples
    12 mm stem spear type pH electrode, glass bodied for measurement of soil and slurry samples
    pH
    0...80 °C
    Epoxy
    12×120 mm
    BNC
    Jenway® pH meters
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.600,00 kr
    1.600,00 kr
    1 * 1 Items
       1.600,00 kr
    Specifikationer:
    New Product
    More
    Specifications
    Cat. No.
    JENW79000-71
    Model
    pH
    Kabellængde
    1 m
    pH-område
    0 - 14 pH
    Reference electrolyte
    AgCl, liquid filled single junction