Static ionizer, ION-100A
ION-100A stand-alone ioniser, 280×150×210 mm, 1,2 kg
Katalog # 611-3318
Leverandør: OHAUS
Gå til:
Product Details & Documents
Electrostatic charges on weighing containers and samples can result in false readings or instability as they create attractive and repulsive forces. The ION-100A is designed to be used in conjunction with an OHAUS balance to neutralise electrostatic charges that can lead to inaccuracies.
- Increases the accuracy of lab measurements
- Fan-free technology neutralises static with no sample disruption
- Compact and versatile design; small footprint
- Durable electrodes can be easily replaced
The ionizer neutralises the electrostatic charge between the test tube and the balance, increasing the accuracy of measurements - particularly with low-volume samples.
Specifikationer
- Vægt:1,2 kg
- Beskrivelse:ION-100A stand-alone ioniser
- Seq:50
- B×D×H:280×150×210 mm