Orders LinkContactUsLinkComponent
sparQ mRNA-Seq Kit
sparQ mRNA-Seq Kit
New Product
Leverandør:   Quantabio
sparQ mRNA-Seq Kit
Leverandør:  Quantabio
New Product
Certifikater
sparQ mRNA-Seq Kit
sparQ mRNA-Seq Kit
2 tilgængelige muligheder nedenfor
Product Details & Documents
Ultra-sensitive mRNA library prep workflow with efficient poly(A) capture.
  • Highly sensitive mRNA detection with input as low as 5 ng of total RNA
  • Reproducible transcript profiling
  • High quality directional RNA library prep with streamlined workflow of 4.75 hours
  • Efficient isolation of mRNA allowing detection of most coding genes
sparQ mRNA-Seq Kit enables efficient preparation of stranded mRNA-seq libraries for Illumina® NGS platforms. The streamlined workflow reduces hands-on time and allows sample to sequencing in a single day. High-quality, reliable mRNA-seq libraries are delivered with consistency and reduced bias from as little as 5 ng and up to 1 μg of total RNA.
Emballering: Store Box 1 in a constant temperature refrigerator at 2°C to 8°C upon receipt. Store Box 2 in a constant temperature freezer at -25°C to -15°C upon receipt.
Product Family Options
Produktinformation
  • No. of reactions
    Tilgængelighed
    Price
  • QUNT95218-024
    sparQ mRNA-Seq kit
    sparQ mRNA-Seq kit
    24
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 6.780,00 kr
    6.780,00 kr
    1 * 1 Each
       6.780,00 kr
    Specifikationer:
    New Product
    New Product
  • QUNT95218-096
    sparQ mRNA-Seq kit
    sparQ mRNA-Seq kit
    96
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 22.970,00 kr
    22.970,00 kr
    1 * 1 Each
       22.970,00 kr
    Specifikationer:
    New Product