2100 Bioanalyzer Instrument
Sizing, quantification, and sample quality control of DNA, RNA, and proteins on a single platform.
- Replaceable electrode cartridges that can be easily exchanged for contamination-free switching between methods
- The system features minimum exposure to hazardous materials to ensure operator safety
- The 2100 Bioanalyzer instrument is compliant with European RoHS directives
- For GLP/GMP environments, the 2100 Bioanalyzer system offers IQ and OQ services, as well as features to enable 21 CF Part 11 compliance
The 2100 Bioanalyzer system is an established automated electrophoresis tool for the sample quality control of biomolecules. The 2100 Bioanalyzer instrument, together with the 2100 Expert Software and Bioanalyzer assays, provide highly precise analytical evaluation of various samples types in many workflows, including next generation sequencing (NGS), gene expression, biopharmaceutical, and gene editing research.
Digital data is provided in a timely manner and delivers objective assessment of sizing, quantitation, integrity and purity from DNA, RNA, and proteins. Minimal sample volumes are required for an accurate result, and the data may be exported in a many different formats for ease-of-use.
: For research use only. Not for use in diagnostic procedures.
- Beskrivelse:2100 Bioanalyzer instrument
- Details:Includes the Bioanalyzer instrument (Model G2939B), electrode cartridge, 2100 Expert software, licenses, priming station, vortexer, test chips, and services