Orders LinkContactUsLinkComponent
Genomic DNA 165 kb Kit
Genomic DNA 165 kb Kit
gDNA 165 kb analysis kit, Femto pulse system kit for the qualitative analysis of HMW gDNA samples from 1300 bp - 165 kb, includes reagents necessary for running 275 samples
Katalog # AGLSFP-1002-0275
Leverandør:   AGILENT
Genomic DNA 165 kb Kit
Katalog # AGLSFP-1002-0275
Leverandør:  AGILENT
Genomic DNA 165 kb Kit
gDNA 165 kb analysis kit, Femto pulse system kit for the qualitative analysis of HMW gDNA samples from 1300 bp - 165 kb, includes reagents necessary for running 275 samples
Price
12.719,65 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Quality control of high molecular weight gDNA and long read NGS libraries.
  • Reliable sizing - Accurate and precise sizing of HMW gDNA samples ranging from 1300 bp to 165 kb
  • Reduced time to results - Optimised pulsed-field separation methods decrease QC analysis time and eliminates the need for overnight PFGE separations
  • Quality metrics - Genomic Quality Number (GQN) quickly scores the integrity of HMW gDNA
  • Ultra-sensitive detection - Input concentrations as low as 5 pg/µl
  • Low sample volume - Only 2 µl required, conserving sample for downstream analysis
The genomic DNA 165 kb kit is used in the assessment of high molecular weight (HMW) genomic DNA quality control required for long-read NGS libraries. This kit is designed to run with an optimised pulsed-field separation method enabling accurate sizing of HMW gDNA through 165 kb.
Long-read NGS library preparations require several gDNA quality control steps. Compared to standard 16-hour PFGE protocols, the Genomic DNA 165 kb kit produces results in as little as 1,5 hours. Replacing traditional PFGE with the Genomic DNA 165 kb kit eliminates days from the tedious library workflow.
Advarsel: For research use only. Not for use in diagnostic procedures.
Specifikationer
  • Beskrivelse:
    gDNA 165 kb analysis kit
  • Details:
    Femto pulse system kit for the qualitative analysis of HMW gDNA samples from 1300 bp - 165 kb, includes reagents necessary for running 275 samples