Orders LinkContactUsLinkComponent
Femto Pulse System
Femto Pulse System
Leverandør:   AGILENT
Femto Pulse System
Leverandør:  AGILENT
Certifikater
Femto Pulse System
6 tilgængelige muligheder nedenfor
Product Details & Documents
Pulsed-field capillary electrophoresis system for QC of high molecular weight DNA and RNA samples.
  • Large fragment resolution - separate high molecular weight DNA smears and fragments through 165 kb in about 1,5 hours
  • Femtogram level sensitivity - achieve 10 times higher sensitivity for nucleic acid smears and up to 100 times higher for nucleic acid fragments
  • Simple sample preparation - minimise sample degradation with fewer pipetting steps and stable reagents
  • Powerful analysis software - one intuitive software program automates analysis of diverse sample types
  • Conserve precious sample - unparalleled sensitivity allows you to use less precious sample and achieve superior detection
  • Make workflows more efficient - eliminate pulsed-field gel electrophoresis from long-read NGS library preparation and BAC analysis
  • Desktop PC supplied with a comprehensive 3-year warranty through HP
By using a pulsed-field power supply, large molecular weight DNA fragments can be separated and nucleic acids detected into the femtogram range. Compared to other non-denaturing gel-based analytical instruments, the Femto Pulse system easily achieves 10 times higher sensitivity for nucleic acid smears and up to 100 times higher sensitivity for nucleic acid fragments.
The ability to separate high molecular weight DNA up to 165 kb and detect nucleic acids down to 50 fg/µl input concentration make the Femto Pulse system ideal for long-read NGS QC, gDNA, small RNA, or cfDNA analysis from low concentration samples.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Capillary array
Produktinformation
  • Details
    Tilgængelighed
    Price
  • AGLSM5330AA-001
    Femto pulse 12-capillary array
    Femto pulse 12-capillary array
    For the Femto Pulse system
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
Inlet buffers
Produktinformation
  • Details
    Tilgængelighed
    Price
  • AGLSDNF-325-0010
    Inlet buffer 5×, 10 ml, for use with Femto pulse system
    Inlet buffer 5×, 10 ml, for use with Femto pulse system
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 185,29 kr
    185,29 kr
    1 * 1 Items
       185,29 kr
  • AGLSDNF-325-0075
    Inlet buffer 5X, 75 ml, for use with Femto pulse system
    Inlet buffer 5X, 75 ml, for use with Femto pulse system
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 75 mL
    1 * 75 mL - 499,76 kr
    499,76 kr
    1 * 75 mL
       499,76 kr
Femto pulse verification kit
Produktinformation
  • Details
    Tilgængelighed
    Price
  • AGLSFP-OQ
    Femto pulse verification kit
    Femto pulse verification kit
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 7.400,12 kr
    7.400,12 kr
    1 * 1 Items
       7.400,12 kr
Reagents
Produktinformation
  • Details
    Tilgængelighed
    Price
  • AGLSGP-435-0100
    Storage solution, 100 ml
    Storage solution, 100 ml
    For the placement of capillaries between runs, For use with the Femto Pulse and Oligo Pro II system
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 554,82 kr
    554,82 kr
    1 * 1 Items
       554,82 kr
Produktinformation
  • Details
    Tilgængelighed
    Price
  • AGLSM5330AA
    Femto pulse system
    Femto pulse system
    Parallel capillary electrophoresis instrument for analysis of nucleic acids, compatible with the Femto Pulse 12-capillary array
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items
Capillary array
Produktinformation
  • Katalog #
    Beskrivelse
    Details
    Tilgængelighed
    Price
  • Femto pulse 12-capillary array
    For the Femto Pulse system
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 22.049,00 kr
    22.049,00 kr
    1 * 1 Items
       22.049,00 kr
Accessories
Produktinformation
  • Katalog #
    Details
    Tilgængelighed
    Price
  • 96 well trays for buffer and waste locations, 50 per pack
    96 well trays for buffer and waste locations, 50 per pack
    AGLSP60-20
    For use with the fragment analyser, Femto Pulse, and ZAG DNA analyser systems
    Log ind for tilgængelighed
    1 * 50 Items - 3.196,59 kr
Produktinformation
  • Katalog #
    Details
    Tilgængelighed
    Price
  • Large open waste tray/tub
    Large open waste tray/tub
    AGLSP60-22
    For use with the fragment analyser, Femto Pulse, and ZAG DNA analyser systems
    Log ind for tilgængelighed
    1 * 1 Items - 166,24 kr