Centrifugal power device with experimental trolley
765-1427
- Beskrivelse:Drive belt for electric motor
- Pakn:1
With the equipment, centrifugal forces that arise during a body's rotation around a specific axis can be measured and examined. The device consists of a plane with a path for a cart. On the underside there is a vertical shaft that is mounted in a bearing. The plane is made to rotate with a laboratory motor connected with a drive belt to the bearing. The experimental trolley is placed on the track and prevented from leaving the track by two bent steel wires. A wire is connected to the trolley that runs over a pulley and on to a dynamometer.