- B×D×H udv.:747×769×1684 mm
- Kapacitet:558/441 L (gross/net)
- Beskrivelse:Refrigerator, SRFfg 5501
- Seq:03
- Model:SRFfg 5501
- Elektriske specifikationer:220 - 240V~, 2,0 A
- Sound level:49 dB(A)
- Kølemiddel:R 600a
- Adgangsporte:1ר 10 mm
- Alarm:Visual and audible
- Afrimning:Automatic
- Display:Monochrome display, touch buttons
- Dørlås:mechanical
- Dørtype:steel
- Hylder/skuffer:5 adjustable, glass shelves
- Vægt:92 (gross)/80 (net) kg
- Pakn:1
- B×D×H indv.:600×537×1460 mm
- Temperatur:3...16 °C (ambient 10…35 °C)
The refrigerators with spark-free interior are easy to clean and the robust glass shelves are simple to remove at 90° door opening. They are super-efficient, utilising the energy-savings potential of every component.
- Alarm relay, alarm test function and custom alarm settings possible
- Min./max. temperature log, self-closing door at <90° and mechanical lock provided
- Retrofittable optional LAN/WiFi, Local API (optional via SmartModule)
- SmartModule can be retrofitted
- Antimicrobial handle with opening mechanism
- Super-efficient energy consumption and forced-air cooling system
- Internal liner material made of commercial grade moulded polystyrol
Temperature fluctuations must be minimal to ensure that the quality of sensitive substances is preserved. These have an impressive maximum deviation of just ±3 °C from the set temperature. This not only maintains the storage climate, but also the value of stored goods. Refrigerators ±3 K deviation from setpoint and with calibration option.
If the temperature falls below +2 °C due to a malfunction, the SafetyDevice takes control, stabilising the temperature, activating the alarm and protecting the contents. Power failure alarm when mains power returns.
Features: Individually adjustable maintenance timer, castors/adjustable feet as accessories. Steel/white external cabinet finish.