Orders LinkContactUsLinkComponent
Avantor sætter videnskaben i gang for en bedre verden | Avantor
Caution label, English, gul/sort, 25×50 mm
Katalog # 116-0952
Leverandør:   DESCO
Katalog # 116-0952
Leverandør:  DESCO
undefined
Caution label, English, gul/sort, 25×50 mm
Price
211,00 kr
Log ind for tilgængelighed
1 * 1 ROL
Product Details & Documents
Caution labels made of white woodfree machine coated midgloss paper.
  • Marked with ESD susceptibility symbol
IEC 61340-5 requires all ESD sensitive devices and assemblies to be labelled with the 'susceptibility' symbol. It is intended to identify devices and assemblies that are susceptible to ESD. Mentioned on individual components and related documents to state "this device is static sensitive: do not touch without approrpiate precautions"
Certifications: Compliant with IEC 61340-5-1.
Delivery information: Supplied in rolls of 1000 individual labels.
Specifikationer
  • Farve:
    gul/sort
  • Beskrivelse:
    Caution label, English
  • Dimensioner:
    25×50 mm