Interconnect mat cord, SCS
Interconnect mat cord, with 10 mm male stud snap fasteners, sort, 10'
Catalog # 115-3047
Supplier: DESCO
Jump to:
Product Details & Documents
Daisy-chains two worksurface mats for the monitoring of two workstations with the 724 workstation monitor.
- Reduces inadvertent snagging that may disconnect the cord from the worksurface mat
- No resistor
This monitor/table mat interconnect cord is a straight cord with male stud snap fasteners at each end, and no resistors.
Certifications: Meets ANSI/ESD S20.20 and Grounding standard ANSI/ESD S6.1.
Specifications
- Farve:sort
- Beskrivelse:Interconnect mat cord, with 10 mm male stud snap fasteners
- Længde:10'