Orders LinkContactUsLinkComponent
Cleaning and Storage Solutions
Cleaning and Storage Solutions
Opløsning til elektrodeopbevaring, 500 ml
Katalog # 662-9223
Leverandør:   HAMILTON BONADUZ
Cleaning and Storage Solutions
Katalog # 662-9223
Leverandør:  HAMILTON BONADUZ
CAS nummer:  
Cleaning and Storage Solutions
Cleaning and Storage Solutions
Opløsning til elektrodeopbevaring, 500 ml
CAS nummer:  
Price
275,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Hamilton storage solution is recommended to increase sensor longevity when not in use. The liquid solution is acid buffered to help with sensor response time after storage as well as prevent any biological growth.
Documents
Specifikationer
Specifikationer
  • Pakn:
    500 ml
  • Beskrivelse:
    Opløsning til elektrodeopbevaring