2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Isopropylalkohol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:82 °C (1013 hPa)
- Melting Pt:–89 °C
- Density:0,786 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Stue temperatur
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
2 Options Available Below
Product Details & Documents
Synonyms: Isopropylalkohol
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:82 °C (1013 hPa)
- Melting Pt:–89 °C
- Density:0,786 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Stue temperatur
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
Documents
Specifications for Product Family
Specification Test Results
- UV Absorbance - 260 - 400 nm:max. 0,01
- UV Absorbance - 230 nm:max. 0,15
- UV Absorbance - 220 nm:max. 0,30
- Assay (CH3CHOH CH3):min. 99,8 %
- Residue after Evaporation:max. 1,0000 ppm
- Water (H2O):max. 0,06 %
- LC/MS Suitability - Largest Response on ESI - Positive Mode (as Reserpine):max. 50 ppb
- Trace Impurities - Aluminum (Al):max. 50,0 ppb
- Trace Impurities - * Calcium (Ca):max. 50,0 ppb
- Trace Impurities - Iron (Fe):max. 50,0 ppb
- Trace Impurities - Magnesium (Mg):max. 50,0 ppb
- Trace Impurities - Potassium (K):max. 50,0 ppb
- Trace Impurities - * Sodium (Na):max. 50,0 ppb
Specifications
Specifications
- Catalog No:
- 9827.1000
- 9827.2500
- Environmentally Preferable:
- Sikrere sundhed for mennesker og miljø
- Sikrere sundhed for mennesker og miljø
- Miljøcertificering:
- Chem21 Solvent Selection Guide recommended by default and after discussion
- EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Product Family Options
Product Information
- PaknAvailabilityPrice
- Specifications:
Sikrere sundhed for mennesker og miljø More
Specifications
Cat. No.9827.1000Environmentally PreferableSikrere sundhed for mennesker og miljøMiljøcertificeringChem21 Solvent Selection Guide recommended by default and after discussionSikrere sundhed for mennesker og miljø - Specifications:
Sikrere sundhed for mennesker og miljø More
Specifications
Cat. No.9827.2500Environmentally PreferableSikrere sundhed for mennesker og miljøMiljøcertificeringEPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussionSikrere sundhed for mennesker og miljø