Fibronectin 96-well clear flat bottom TC-treated microplates, Corning® BioCoat®
734-0126
: Corning
- Ant. brønde:96
- Pakket:5 plates
- Steril:-
- Farve:klar
- Beskrivelse:96-well plates
- Seq:03
- Environmentally Preferable:Lav fremstillingspåvirkning
- Miljøcertificering:ISO 14001 - Bedford
- Dyrkningsområde:0,32 cm²
- Pakn:5