Starter kit, Lambda™ Plus, Corning
Lambda™ Plus-startsæt, inkl. fire pipetter og tre rack med pipettespidser
Katalog # CORN4069
Leverandør: Corning
Starter kit, Lambda™ Plus, Corning
Katalog # CORN4069
Leverandør: Corning
Gå til:
Product Details & Documents
The Lambda plus starter kit is designed for laboratories which are about to replace their old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes.
The Lambda plus starter kit contains four Lambda Plus Single-Channel pipettors conveniently packaged with useful accessories (manual, calibration key, colour-coded rings, tube of piston grease and identification stickers).
: This starter kit includes four Lambda Plus Single Channel pipettors in these volume sizes 0,5 to 10 µl, 2,0 to 20 µl, 20 to 200 µl, and 100 to 1000 µl and three racks of Corning IsoTip™ filtered pipette tips for the above sizes.
Specifikationer
- Beskrivelse:Lambda™ Plus-startsæt, inkl. fire pipetter og tre rack med pipettespidser
- Cat. No.:CORN4069
- Miljøcertificering:I-RECs, ISO 14001 - Warsaw
- Environmentally Preferable:Lav fremstillingspåvirkning