Cleanroom Overalls, CleanMax®
Om denne vare
Clean manufactured disposable coveralls are individually packaged and folded to promote easier gowning with less excursions.
- Clean manufactured in silicone-free environment
- Latex-free
- Tunnelled elastic on wrists with thumb loops
- Zipper closure
- For critical environments
- Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.
7 tilgængelige muligheder nedenfor
Product Details & Documents
Clean manufactured disposable coveralls are individually packaged and folded to promote easier gowning with less excursions.
- Clean manufactured in silicone-free environment
- Latex-free
- Tunnelled elastic on wrists with thumb loops
- Zipper closure
- For critical environments
- Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.
Specifications for Product Family
Specifikationer
- Catalog No:
- 114-9589
- 114-9588
- 114-9590
- 114-9584
- 114-9585
- 114-9586
- 114-9587
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Renrum klasse:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Outfit applikationer:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
Specifikationer
Product Family Options
Produktinformation
- FarveTøj størrelseTilgængelighedPrice
- 114-9589Clean manufactured coverallSpecifikationer:
More
Specifications
Cat. No.114-9589LukkestørrelseMed lynlåsRenrum klasse100/ISO 5Outfit applikationerCleanroom - 114-9588Clean manufactured coverallSpecifikationer:
More
Specifications
Cat. No.114-9588LukkestørrelseMed lynlåsRenrum klasse100/ISO 5Outfit applikationerCleanroom - 114-9590Clean manufactured coverallSpecifikationer:
More
Specifications
Cat. No.114-9590LukkestørrelseMed lynlåsRenrum klasse100/ISO 5Outfit applikationerCleanroom - 114-9584Clean manufactured coverallSpecifikationer:
More
Specifications
Cat. No.114-9584LukkestørrelseMed lynlåsRenrum klasse100/ISO 5Outfit applikationerCleanroom - 114-9585Clean manufactured coverallSpecifikationer:
More
Specifications
Cat. No.114-9585LukkestørrelseMed lynlåsRenrum klasse100/ISO 5Outfit applikationerCleanroom - 114-9586Clean manufactured coverallSpecifikationer:
More
Specifications
Cat. No.114-9586LukkestørrelseMed lynlåsRenrum klasse100/ISO 5Outfit applikationerCleanroom - 114-9587Clean manufactured coverallSpecifikationer:
More
Specifications
Cat. No.114-9587LukkestørrelseMed lynlåsRenrum klasse100/ISO 5Outfit applikationerCleanroom