Orders LinkContactUsLinkComponent
Ionic strength adjustor (ISA)
Ionic strength adjustor (ISA)
500 mL
Katalog # HACH2488349
Leverandør:   Hach
Ionic strength adjustor (ISA)
Katalog # HACH2488349
Leverandør:  Hach
CAS nummer:  
Ionic strength adjustor (ISA)
Ionic strength adjustor (ISA)
500 mL
CAS nummer:  
Price
230,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Nitrate Ionic Strength Adjustor (ISA) Buffer Solution for sample treatment in nitrate determination by ion selective electrode.
Specifikationer
Specifikationer
  • Pakn:
    500 mL
Specifikationer