Manual dispenser and applicator, Scotch-Weld™ EPX™
Catalog # MMMAEPX400H
Supplier: 3M
Specifications
- Volumen:400 ml
- Farve:grå
- Beskrivelse:Applicator
- Pakn:1
Specifications
About this item
Manual applicator for dispensing EPX adhesive in 38 and 50 ml cartridges supplied with two plungers for 1:1/2:1 and 10:1 adhesive.
- Dispenses two-component 3M™ Scotch-Weld™ structural adhesive
- Offers take-it-to-the-job convenience without the need for air hoses or electrical cords
- Ensures correct mix ratio during dispensing; no need to weigh components
- Minimises waste and mess associated with hand mixing
- Reduce plastic waste
Dispense 3M™ Scotch-Weld™ adhesive in duo-pack cartridges with 3M™ Scotch-Weld™ EPX manual applicator. Our applicator mixes and applies two part adhesives. It is commonly used in industrial, metalworking and transportation applications.
Use with both 1:1 and 2:1 200 ml cartridges.